Trifluoreddiksyre

Leverandør: Thermo Fisher Scientific
Danger

28903 28904 28901 28902
PIER28903EA 2800 NOK
PIER28903 PIER28904 PIER28901 PIER28902
Trifluoreddiksyre
Trifluoreddiksyre
Formel: C₂HF₃O₂
Kokepunkt: 71,8 °C (1013 hPa)
Smeltepunkt: –15 °C
Tetthet: 1,490 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004169
CAS-nummer: 76-05-1
EINECS: 200-929-3
FN: 2699
ADR: 8,I
Merck Indeks: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR