Maursyre ( metansyre) ≥99%

Leverandør: Thermo Fisher Scientific
Danger

28905
PIER28905EA 1525 NOK
PIER28905
Maursyre ( metansyre) ≥99%
Maursyre ( metansyre)
Formel: HCO₂H
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003297
CAS-nummer: 64-18-6
EINECS: 200-579-1
FN: 1779
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR