D(+)-Biotin

Leverandør: Thermo Fisher Scientific

Synonymer: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

29130
PIER29130EA 2700 NOK
PIER29130
D(+)-Biotin
D(+)-Biotin
Formel: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Smeltepunkt: 232…233 °C
Tetthet: 1.41 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005541
CAS-nummer: 58-85-5
EINECS: 200-399-3
Merck Indeks: 13,01231

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR