TMEA (Tris(2-maleimidoethyl)amine), Pierce™

Leverandør: Thermo Fisher Scientific
Warning

33043
PIER33043EA 3325 NOK
PIER33043
TMEA (Tris(2-maleimidoethyl)amine), Pierce™
TMEA (Tris(2-maleimidoethyl)amine)

Thermo Scientific Pierce TMEA is a trifunctional crosslinker containing three reactive maleimide groups to enable three-way conjugation and polymerization of sulfhydryl-containing molecules.


  • Reactive groups: maleimide (on three termini)
  • Reactive towards: sulfhydryl groups
  • 3-way, sulfhydryl-to-sulfhydryl crosslinker; tris (2-maleimidoethyl) amine, composed of three maleimide groups on short spacer arms
  • Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
  • Noncleavable
  • Useful for the preparation of trimeric complexes of cysteine-containing peptides and other sulfhydryl-containing compounds
  • Core molecule for the construction of dendritic polymers


NB: For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₈H₁₈N₄O₆
Lagringstemperatur: Kjøleskap
CAS-nummer: 139112-38-2

Bestill nå

Spesifikasjon Testresultater

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homotrifunctional
Reactive Moiety Maleimide
Spacer Mid-Length
Form Solid
Quantity 50 mg
Type Crosslinker
Solubility DMF, DMSO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR