DAB substrate
Leverandør: Thermo Fisher Scientific
34002
PIER34002EA 1625 NOK
PIER34002
DAB substrate
Elektroforese reagenser Western Blotting Reagents
DAB (3,3'-diaminodbenzidine) is a sensitive colorimetric substrate used with horseradish peroxidase (HRP) in immunohistochemistry and immunoblotting applications. HRP catalyses hydrogen peroxide oxidation of substrates such as DAB. Electrons are transferred by HRP from the DAB to the peroxide to yield an insoluble brown product.
Leveringsinformasjon: Kit contains DAB Solution (10X), 25 ml; Stable Peroxide Substrate Buffer, 250 ml.