Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Leverandør: Thermo Fisher Scientific
34035
PIER34035EA 2450 NOK
PIER34035
Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Elektroforese reagenser Western Blotting Reagents
NBT (nitro-blue tetrazolium chloride) yields a blue-purple, water-insoluble product upon reduction of the tetrazolium salt. The product of reduction is NBT-formazan. This substrate is widely used for immunochemistry applications because the color is linear and stable. NBT is used as the substrate for both glucose oxidase and alkaline phosphatase systems.