Film, CL-XPosure™
Leverandør: Thermo Fisher Scientific
34090 34089
PIER34090EA 2825 NOK
PIER34090 PIER34089
Film, CL-XPosure™
Autoradiografifilm Filmer
CL-XPosure™ Film is a clear blue X-ray film recommended for the detection and documentation of blotting assays.
  • High performance - just as sensitive as other commercially available autoradiography films
  • Economical - achieve high-quality images at an affordable price
  • Convenient - available in five sheet sizes in packages containing 100 non-interleaved sheets
  • Perfect match - validated for use with SuperSignal chemiluminescent substrates and Pierce ECL and ECL Plus chemiluminescent substrates
CL-XPosure™ works with chemiluminescent horseradish peroxidaseMore Product Information