Fast western blot kit, SuperSignal™ West Dura, Pierce™
Leverandør: Thermo Fisher Scientific
35070 35071
PIER35070EA 6400 NOK
PIER35070 PIER35071
Fast western blot kit, SuperSignal™ West Dura, Pierce™
Elektroforese reagenser Western Blotting Reagents
The Fast Western Blot Kits, SuperSignal™ West Dura provide reagents and protocol for reliable and rapid, 1 hour chemiluminescent Western blotting at femtogram levels of sensitivity. These complete Fast Western Blot Kits requires minimal hands on time and yield results comparable to classical Western blotting with SuperSignal™ West Dura Chemiluminescent Substrate. This substrate produces a signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed. The complete kit contains all the reagents necessary to block and probe a Western blot with mouse or rabbit primary antibody.
  • Fast - femtogram levels of detection in about 1 hour
  • No expensive hardware or vacuum required; no clogging issues
  • Optimised protocol makes Western blot analysis easier
  • No expensive consumables or extra equipment required
  • Kit stable for 1 year stored at 4 °C
The Fast Western Blot Kit protocol is a quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other rapid Western blotting systems. These complete Pierce Fast Western Blot Kits require minimal hands-on time and yield results comparable to classical Western blotting with SuperSignal West Dura Chemiluminescent Substrate. Like other Western blotting methods, the included SuperSignal West Dura Substrate produces a signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed. The complete kit contains all the reagents necessary to block and probe a Western blot with mouse primary antibody.
Leveringsinformasjon: Kit includes Fast Western antibody diluent, Fast Western 10X wash buffer, Fast Western (mouse or rabbit) optimised HRP reagent, SuperSignal™ West Dura luminol/enhancer solution, SuperSignal™ West stable peroxide solution.
NB: For research use only. Not for use in diagnostic procedures.