FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)

Supplier: Thermo Fisher Scientific
Warning

46424
PIER46424EA 3050 NOK
PIER46424
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
FITC (Fluorescein 5-isothiocyanate, Fluorescein isothiocyanate isomer I)
Formula: C₂₁H₁₁NO₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00005063
CAS Number: 3326-32-7
EINECS: 222-042-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR