myImageAnalysis™ software, Pierce™
Leverandør: Thermo Fisher Scientific
62237
PIER62237EA 37325 NOK
PIER62237
myImageAnalysis™ software, Pierce™
Programvare Laboratoriesoftware
myImageAnalysis™ Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.
  • Intuitive design
  • Load images in popular file formats, analyze images from nearly any source, and easily share the results
  • Open two to six image files at once to review them simultaneously or toggle between them to edit them in parallel
myImageAnalysis™ software comprises a complete set of easy-to-use research tools for the quantitative digitalMore Product Information