Pierce™, Biodyne™ A Blotting Membranes, Nylon
Leverandør: Thermo Fisher Scientific
77015
PIER77015EA 6525 NOK
PIER77015
Pierce™, Biodyne™ A Blotting Membranes, Nylon
Overføringsmembraner
Biodyne™ A nylon membranes are sheets of unmodified nylon that provide excellent binding characteristics for nucleic acid transfer, blotting and detection methods. Biodyne™ A nylon membrane is manufactured with a matrix pore size of 0,45 micron. It contains both negatively and positively charged groups, enabling it to bind many kinds of macromolecules.
  • Versatile and durable - achieve strong DNA binding by baking or UV-crosslinking without damaging the membrane
  • Pre-cut 8×12 cm sheets are the ideal size for standard mini-gel transfers and microplate dot-