Blotting membranes, nylon, Biodyne™ B
Leverandør: Thermo Fisher Scientific
77016
PIER77016EA 4525 NOK
PIER77016
Blotting membranes, nylon, Biodyne™ B
Overføringsmembraner
Biodyne™ B nylon membranes are sheets of positively charged nylon that provide excellent binding characteristics for nucleic acid transfer, blotting and detection methods. Biodyne™ B nylon membrane is manufactured with a matrix pore size of 0,4 micron. It contains exclusively positive charges, providing the highest possible binding capacity for negatively charged molecules, like DNA and RNA.
  • Versatile and durable - achieve strong DNA binding by baking or UV-crosslinking without damaging the membrane
  • Pre-cut 8×12 cm sheets are the ideal size for standard