Y-PER™ plus Yeast Protein Extraction Reagent, Dialysable
Leverandør: Thermo Fisher Scientific
78999
PIER78999EA 5650 NOK
PIER78999
Y-PER™ plus Yeast Protein Extraction Reagent, Dialysable
Buffere Buffere, ekstrasjons
This extraction reagent is formulated to effectively lyse yeast and bacterial cells to extract and solubilise proteins for analysis.
  • No reducing or chelating agents
  • Designed for room temperature storage
  • Fast lysis completes after 20 minutes hands-off incubation
  • Requires no rigorous mechanical disruption or lengthy enzymatic treatments
The reagent is used for Western blots, GST and histidine-tagged purification based applications. This fully dialysable formulation, allows complete exchange of the extracted protein into any buffer required by downstream techniques. The tris-based buffer has a very low ionic strength and does not include sodium chloride. The reagent is compatible with both the Thermo Scientific Pierce BCA and Coomassie Plus Protein Assays.