ESI negative ion calibration solution, LTQ, Pierce™
Leverandør: Thermo Fisher Scientific
88324
PIER88324EA 3350 NOK
PIER88324
ESI negative ion calibration solution, LTQ, Pierce™
Standarder Fargereferanse og spekroskopistandarer
Ion calibration solution are mixtures of highly purified ionizable molecules specifically designed for negative mode calibration of ion trap and orbitrap instruments.
  • Mass spectrometry grade reagents in a non-leachable container
  • Load the mass reference standard into a syringe and inject into the instrument
  • Strong peaks—mixture of SDS, sodium taurocholate and Ultramark 1621 in a acetonitrile/methanol/acetic solution.
It is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific mass spectrometers.