Protein transfection reagent, Pierce™
Leverandør: Thermo Fisher Scientific
89850
PIER89850EA 6025 NOK
PIER89850
Protein transfection reagent, Pierce™
Transfeksjonsreagenser
This Pierce™ protein transfection reagent is a cationic lipid mixture for complexation with proteins, peptides, antibodies and other biologically active molecules to allow their direct intracellular delivery.
  • No need to isolate, clone and transfect gene sequences thu saving time
  • Optimum delivery in three to four hours after incubation
  • Delivery complex is non-covalent with proteins and peptides retaining biological activity after delivery
  • Works with proteins, peptides and fluorescently labeled antibodies making it versatile
Molecules that have been successfully delivered with the transfection reagent include β-galactosidase, fluorescent-antibody, green fluorescent protein (GFP), high and low molecular weight dextran sulfate, phycoerythrin-BSA, caspase 3, caspase 8 and granzyme B.