L(+)-Lysine dihydrochloride for SILAC

Leverandør: Thermo Fisher Scientific

89987
PIER89987EA 580 NOK
PIER89987
L(+)-Lysine dihydrochloride for SILAC
L(+)-Lysine dihydrochloride
Formel: C₆H₁₆Cl₂N₂O₂
MDL nummer: MFCD00066389
CAS-nummer: 657-26-1
EINECS: 211-518-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR