Protein precipitation plates, Pierce™
Leverandør: Thermo Fisher Scientific
90036 90037
PIER90036EA 3100 NOK
PIER90036 PIER90037
Protein precipitation plates, Pierce™
Mikroplater Filterplater
Pierce™ protein precipitation plates provide an easy, 96-well format for simple, rapid and automatable precipitation and removal of proteins from plasma, serum and other biological fluid samples. The plates are the ideal choice for downstream processing of plasma protein binding assay samples prepared with equilibrium dialysis systems.
  • Prepares small molecule samples for HPLC or LC-MS in 10 to 30 minutes
  • No acetonitrile leakage for up to four hours
  • Leach-free graded hydrophobic frit prevents blockage
  • Filtrates are easily collected by vacuum, positive