Protein quantitation reagent, TMTzero™ label reagent
Leverandør: Thermo Fisher Scientific
90067
PIER90067EA 2575 NOK
PIER90067
Protein quantitation reagent, TMTzero™ label reagent
Proteinekstraksjons Reagenser for proteinekstraksjon
Pierce™ TMTzero™ Label Reagent serves as a "light" tag for MS-level peptide quantitation similar to duplex isotopic metabolic labeling, such as SILAC, or isotopic dimethylation labeling.