Rapid equilbrium dialysis (RED) device, Pierce™
Leverandør: Thermo Fisher Scientific
89811 90006 90007 90085 90087 90088 99006 90112 91012
PIER89811EA 6175 NOK
PIER89811 PIER90006 PIER90007 PIER90085 PIER90087 PIER90088 PIER99006 PIER90112 PIER91012
Rapid equilbrium dialysis (RED) device, Pierce™
Rør Dialyserør
Single-Use RED (rapid equilibrium dialysis) Plate is composed of disposable high-density polypropylene and is pre-loaded with 48 equilibrium dialysis membrane inserts. Each insert is comprised of two side-by-side chambers separated by an O-ring-sealed vertical cylinder of dialysis membrane with varying molecular-weight cutoffs (8 or 12 kD MWCO).
  • Easy and ready to use
  • Design enables equilibrium to reach 200 min⁻¹ agitation with in three to four hours
  • Reproducible and accurate for plasma binding assays
  • Robust compartmentalised design eliminates potential for