DSP (Dithiobis(succinimidyl propionate)), No-Weigh™ Format, Pierce™

Leverandør: Thermo Fisher Scientific

A35393
PIERA35393EA 1825 NOK
PIERA35393
DSP (Dithiobis(succinimidyl propionate)), No-Weigh™ Format, Pierce™
DSP (Dithiobis(succinimidyl propionate))

Thermo Scientific Pierce DSP (Lomant's Reagent) is a water-insoluble, homo-bifunctional N-hydroxysuccimide ester (NHS ester) crosslinker that is thiol-cleavable, primary amine-reactive, and useful for many applications. DSP contains an amine-reactive NHS ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7–9 to form stable amide bonds and releasing N-hydroxy-succinimide. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS ester crosslinking reagents. DSP is non-sulfonated and insoluble in water, so it must first be dissolved in an organic solvent and then added to the aqueous reaction mixture. Because DSP does not possess a charged group, it is lipophilic and membrane-permeable and so useful for intracellular and intramembrane conjugation. A sulfonated analog of DSP (DTTSP) is water soluble. DSS, the non-cleavable analog of the DSP crosslinker is also available for applications that require a stable spacer arm that cannot be cleaved in the presence of reducing agents.


  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Amine-reactive NHS ester reacts rapidly with any primary amine-containing molecule
  • Disulfide bond in the spacer arm is readily cleaved by 10-50 mM DTT or TCEP at pH 8.5
  • Spacer arm also cleavable with reducing SDS-PAGE sample loading buffer
  • Cleavable crosslinker allows separation of crosslinked products
  • Water-insoluble (dissolve first in DMF or DMSO); compare to DTSSP (Sulfo-DSP)
  • Membrane-permeable, allowing for intracellular crosslinking


DSP contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxy-succinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS-ester crosslinking reagents. DSS, the non-cleavable analog of DSP crosslinker is also available for applications that require a stable spacer arm that cannot be cleaved in the presence of reducing agents.


Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.


NB: For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₄H₁₆N₂O₈S₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00042045
CAS-nummer: 57757-57-0

Bestill nå

Spesifikasjon Testresultater

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Cleavable, Long
Form Powder
Quantity 50 mg / 1 g
Type Crosslinker
Solubility DMF, DMSO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR