High Select™ top14 abundant protein depletion resin, Pierce™

Leverandør: Thermo Fisher Scientific

A36372
PIERA36372EA 38350 NOK
PIERA36372
High Select™ top14 abundant protein depletion resin, Pierce™
Proteinrense Protein Purification Kits

Thermo Scientific™ High Select Top14 Abundant Protein Depletion Resin decreases the abundant protein and antibody components of human plasma samples, helping prepare the samples for mass spectrometry or 1D or 2D electrophoresis.


  • Flexible: Provided in bulk format for depletion of user-defined amount of human plasma sample
  • Optimised: Removes >95% of IgG and >95% of albumin
  • Fast: Process samples in ~10 minutes
  • Economical: Cost-effective bulk format


High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilised antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resin is provided in an economical bulk format that may be used to fill columns or plates, depending on sample volumes.


Analysis of human fluids is often complicated by the presence of high concentrations of albumin and IgG that can make up more than 70% of total serum protein. Removal of these proteins is often essential for the study of low-abundant proteins. High Select Top14 Abundant Protein Depletion Resin can deplete greater than 95% of the 14 most abundant proteins in serum. Columns or plates are filled with the resin based on the needs of the user, and samples processed in approximately 10 minutes. The depletion of highly abundant proteins enables the detection of low abundant proteins in samples and subsequent identification by mass spectrometry or 1D or 2D gel electrophoresis.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR