Sulpho-NHS-LC-Biotin, Premium Grade, Pierce™

Leverandør: Thermo Fisher Scientific

Synonymer: Sulphosuccinimidyl-6-(biotinamido)hexanoate, Biotinamidohexanoic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt

PG82075
PIERPG82075EA 4850 NOK
PIERPG82075
Sulpho-NHS-LC-Biotin, Premium Grade, Pierce™
Sulpho-NHS-LC-Biotin

Thermo Scientific Pierce Premium Grade Sulfo-NHS-LC-Biotin is our highest quality formulation of this popular amine-reactive biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount.


  • Sulfo-NHS-LC-Biotin—popular amine-reactive biotinylation reagent for antibody labeling
  • High quality—identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review


Compared to the standard grade product, Premium Grade Sulfo-NHS-LC-Biotin provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents.


NB: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store desiccated at -20° C.

Formel: C₂₀H₂₉N₄NaO₉S₂
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00078534
CAS-nummer: 127062-22-0

Bestill nå

Spesifikasjon Testresultater

Product Type Sulfo-NHS-LC-Biotin
Quantity 100mg/1 g
Product Line Pierce™
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Label Type Biotin & Analogs
Spacer Mid-Length

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR