Eyewash station, Combibox DUO
Leverandør: Plum Safety
4862
PLUM4862EA 1300 NOK
PLUM4862
Eyewash station, Combibox DUO
Førstehjelpsutstyr Øyeskyllestasjon
Wall-mounted eye wash box made from maintenance-free material for quick first aid of eye injuries. The box contains one 500 ml bottle of pH Neutral DUO and one 500 ml bottle of Plum eye wash DUO.
  • Dust-tight wall station in high-impact PS
  • Quick and efficient eye wash
  • Quick and easy to mount on the wall close to the workplace
  • With clear pictograms and mirror
  • Particularly suited for dusty and dirty production areas
W×D×H: 230×110×280 mm