Cleaning swab, PurSwab®
Leverandør: PURITAN MEDICAL
816
PMPC816EA 4100 NOK
PMPC816
Cleaning swab, PurSwab®
Svabere Svabere for rengjøring
Proctoscopic swab made with USP grade rayon fibre that is carded on-site to maintain consistency.
  • Large rayon tip is extra-absorbent
Suitable for use in obstetrics and gynecology preparation and examinations as well as for cell collection in veterinary applications.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904