Mini-tipped swabs
Leverandør: PURITAN MEDICAL
826-WC
PMPC826-WCEA 8050 NOK
PMPC826-WC
Mini-tipped swabs
Svabere Svabere for klinisk bruk
Tapered swabs designed for precision application and hard-to-reach areas.
  • Absorbent soft smooth tip and sturdy wooden handle
  • Double end for twice the use
Suitable for jewelry manufacturing and art restoration.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904