Small cotton swab
Leverandør: PURITAN MEDICAL
868-WCS
PMPC868-WCSEA 6025 NOK
PMPC868-WCS
Small cotton swab
Svabere Svabere for rengjøring
The cotton-tipped applicator is produced with high quality USP grade cotton.
  • Bonded securely to a sturdy handle by an aqueous based adhesive
  • Small tip for hard-to-reach or small areas
  • Carded and coiled onsite to maintain consistency
Suitable for cleaning medical devices, to remove oil or dirt from machine parts, in veterinary care, and the cleaning of sensors or electronic components.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904