Swabs, PurSwab®
Leverandør: PURITAN MEDICAL
870-PCDBL
PMPC870-PCDBLEA 6000 NOK
PMPC870-PCDBL
Swabs, PurSwab®
Svabere Svabere for rengjøring
These double-ended swabs with cotton tips are ideal for cleaning sensitive components in critical environments.
  • Low in NVR
  • Highly absorbent and does not swell in solvents
  • Lint- and residue-free
  • Sharp, double pointed tip with fine paper handle
  • Anti-static packaging contains no silicone
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904