Swabs, PurSwab®, cotton head applicator
Leverandør: PURITAN MEDICAL
896-PC
PMPC896-PCEA 3850 NOK
PMPC896-PC
Swabs, PurSwab®, cotton head applicator
Svabere Svabere for rengjøring
The applicator has a single-ended smooth cylindrical cotton tip on a white paper shaft.
  • Easy disposal
  • Lint-less and residue free
  • Highly absorbent cotton does not swell in solvents
Suitable for cleaning disk drives, optics, PC board assembly and other critical industrial and sensitive components.
Forpakning: 50 in heat sealed poly bag - 1000 per case.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904