Buckets, Rectangular, PP
Leverandør: PFENNIG REINIGUNGSTECHNIK
2000601 2000602 2000605 2000603 2000604
PPSA2000601EA 260 NOK
PPSA2000601 PPSA2000602 PPSA2000605 PPSA2000603 PPSA2000604
Buckets, Rectangular, PP
Bøtter
Standard for Clino® system trolleys, rectangular bucket with handle. Also suitable for the pre-preparation of textiles.
  • With scaling from 1 to 5 L
  • Coloured to facilitate use as part of a colour coded system
  • Good chemical resistance
  • Good temperature resistance