Accessories for Cleaning Trolley Clino® CR-1 EM-CR with EasyMop CR
Leverandør: PFENNIG REINIGUNGSTECHNIK
3500100 3500150 2014030 2014010 2013430 2013440
PPSA3500100EA 1275 NOK
PPSA3500100 PPSA3500150 PPSA2014030 PPSA2014010 PPSA2013430 PPSA2013440
Accessories for Cleaning Trolley Clino® CR-1 EM-CR with EasyMop CR
Traller
These niro steering castors with or without brake are electrically conductive for the use in technical cleanrooms. Also autoclavable special castors for use in sterile areas.
  • Easy to move
  • Running and steering comfort
  • Maximum ergonomics