Telescopic Handle
Leverandør: PFENNIG REINIGUNGSTECHNIK
2016114
PPSA2016114EA 2150 NOK
PPSA2016114
Telescopic Handle
Mopper
Ultra lightweight stainless steel telescopic handle with toggle lever, sleeve and quick-release fastener for Clino®Connect and Clino®Connect Lock.
  • Ultralight and stable
  • Height-adjustable
  • Autoclavable
Areas od application: Technical cleanrooms, life science, pharmaceutical cleanrooms, laboratories