Cleaning trolley Clino®, Profi C7e
Leverandør: PFENNIG REINIGUNGSTECHNIK
2097275
PPSA2097275EA 0 NOK
PPSA2097275
Cleaning trolley Clino®, Profi C7e
Traller
The Clino® Profi C7e trolley system made of stainless steel flat wringer for grey areas.
  • Flat wringer Clino provides defined pressure point for uniform residual moisture
  • Disposal unit can be removed in retirement and is interchangeable and extendable
  • Frame and attachments can be autoclaved
  • Double ball-bearinged guide wheels (125 mm)
Leveringsinformasjon: Supplied with bucket (25 L, 15 L and 3 ×6 L), 3 trays and disposal unit (1 ×70 L).