Cleanroom mops, MicroSicuro trapezoid CR/A
Leverandør: PFENNIG REINIGUNGSTECHNIK
2700293
PPSA2700293EA 245 NOK
PPSA2700293
Cleanroom mops, MicroSicuro trapezoid CR/A
Mopper
Mops for cleaning surfaces like insulators, and cleaning of hard-to-reach areas. Made up of 100% PES microfibre with soft brush special.
  • Removes 99,9% of all particles in a one-step process
  • Safe disinfection of GMP areas by validating wetting behavior
  • With water-permeable cover sheet polyester bag and drainage function lining
  • Suitable to all mop frames Trapez
  • Autoclavable up to 50 preparation cycles, disinfectant proof and resistant to gamma rays
Ideal for removing disinfectant residues and maximum square footage.