Multi-Use Mop for Cleanroom, MultiSicuro PW
Leverandør: PFENNIG REINIGUNGSTECHNIK
3500285
PPSA3500285EA 340 NOK
PPSA3500285
Multi-Use Mop for Cleanroom, MultiSicuro PW
Mopper
Multi-use mop with the weight advantage of a disposable mop.
  • Autoclavable up to 50 reprocessing cycles, disinfectant resistant, gamma radiation resistant
  • High area performance due to absorbent material
  • Minimal own particle release
  • Very high quality, clean room suitable processing
  • Pre-washed ready for use
  • White edging tape on the pockets, pre-washed at 90 °C
Sertifiseringer: IPA "Tested Device" Certification.