Cleanroom Sword, CR/A
Leverandør: PFENNIG REINIGUNGSTECHNIK
3500951
PPSA3500951EA 850 NOK
PPSA3500951
Cleanroom Sword, CR/A
Mopper
Flexible holder for cleaning hard-to-reach places and narrow gaps.
  • Autoclavable
  • ClinoConnect Lock handle
  • Locking mechanism
  • Cleaning of hard-to-reach areas and narrow gaps
Grip material: Polyamide, glass-fibre reinforced
Fin material: Plastic Purell, FE silicon manganese wire