Broom head for cleanrooms
Leverandør: PFENNIG REINIGUNGSTECHNIK
3590100
PPSA3590100EA 755 NOK
PPSA3590100
Broom head for cleanrooms
Koster
Brooms for hygienically sensitive areas in cleanrooms. Material: PP (body), PBT (bristles).
Designed for the removal of dirt and coarse contaminations.