Autoclave, Front-Loading
Leverandør: Priorclave
EH150STK-BASE
PRIOEH150STK-BASEEA 0 NOK
PRIOEH150STK-BASE
Autoclave, Front-Loading
Sterilisatorer Autoklav, dampsterilisering
A front-loading autoclave with a traditional cylindrical vessel is the best option available for research, education, and quality assurance facilities.
  • Configuration: Benchtop, front-loading
  • Head-to-head with comparable conventional rectangular autoclaves
  • Offering similar cycle times with lower maintenance burdens
  • Substantial energy, water, and lifetime cost savings
  • Temperature range: max: 138 °C (280, 4 °F); min: 0 °C (32 °F)
  • Applications: Medical, for research
A cylindrical chamber is better suited to high pressure micro-environments, requires lessMore Product Information