4-Chloropyrocatechol

Leverandør: LGC Standards PROMOCHEM

Synonymer: 4-Chloro-1,2-dihydroxybenzene, 4-Chlorobenzene-1,2-diol, 4-Chlorocatechol

HXC001
PROOHXC001EA 0 NOK
PROOHXC001
4-Chloropyrocatechol
4-Chloropyrocatechol
Formel: C₆H₅ClO₂
MW: 144,56 g/mol
Kokepunkt: 267 °C (1013 hPa)
Smeltepunkt: 90…91 °C
MDL nummer: MFCD00059614
CAS-nummer: 2138-22-9
EINECS: 218-381-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR