Gentamicin sulphate

Leverandør: LGC Standards PROMOCHEM
Danger

ICRS0319
PROOICRS0319EA 0 NOK
PROOICRS0319
Gentamicin sulphate
Gentamicin sulphate
Smeltepunkt: 218…237 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00270181
CAS-nummer: 1405-41-0
EINECS: 215-778-9
Merck Indeks: 13,04403

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR