Tetracycline hydrochloride

Leverandør: LGC Standards PROMOCHEM
Warning

ICRS0429
PROOICRS0429EA 0 NOK
PROOICRS0429
Tetracycline hydrochloride
Tetracycline hydrochloride
Formel: C₂₂H₂₅ClN₂O₈
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00078142
CAS-nummer: 64-75-5
EINECS: 200-593-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR