Kaliumsorbat

Leverandør: LGC Standards PROMOCHEM
Warning

Synonymer: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

LGC7305
PROOLGC7305EA 0 NOK
PROOLGC7305
Kaliumsorbat
Kaliumsorbat
Formel: CH₃CH=CHCH=CHCOOK
MW: 150,22 g/mol
Smeltepunkt: 270 °C
Tetthet: 1,36 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016546
CAS-nummer: 24634-61-5
EINECS: 246-376-1
Merck Indeks: 13,07756
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR