Methotrexate

Leverandør: LGC Standards PROMOCHEM
Danger

Synonymer: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid

MM0040.00
PROOMM0040.00EA 0 NOK
PROOMM0040.00
Methotrexate
Methotrexate
Formel: C₂₀H₂₂N₈O₅
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00150847
CAS-nummer: 59-05-2
EINECS: 200-413-8
FN: 2811
ADR: 6.1,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR