Vinylacetat

Leverandør: LGC Standards PROMOCHEM
Danger

Synonymer: Acetic acid vinyl ester

MM0537.01
PROOMM0537.01EA 0 NOK
PROOMM0537.01
Vinylacetat
Vinylacetat
Formel: C₄H₆O₂
MW: 86,09 g/mol
Kokepunkt: 72,3 °C (1013 hPa)
Smeltepunkt: –100 °C
Tetthet: 0,932 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008713
CAS-nummer: 108-05-4
EINECS: 203-545-4
FN: 1301
ADR: 3,II
Merck Indeks: 12,10130

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR