1,5-Pentamethylene-1H-tetrazole

Leverandør: LGC Standards PROMOCHEM

Synonymer: Pentylenetetrazole

MM0692.00
PROOMM0692.00EA 0 NOK
PROOMM0692.00
1,5-Pentamethylene-1H-tetrazole
1,5-Pentamethylene-1H-tetrazole
Formel: C₆H₁₀N₄
MW: 138,17 g/mol
MDL nummer: MFCD00005939
CAS-nummer: 54-95-5
EINECS: 200-219-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR