HEPES sodium salt

Leverandør: LGC Standards PROMOCHEM

Synonymer: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, HEPES sodium, HEPES Na

NIST-2182
PROONIST-2182EA 0 NOK
PROONIST-2182
HEPES sodium salt
HEPES sodium salt
Formel: C₈H₁₇N₂NaO₄S
MW: 260,29 g/mol
Smeltepunkt: ? 105 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036463
CAS-nummer: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR