Beryllium standard solution
Leverandør: LGC Standards PROMOCHEM
NIST-3105A
PROONIST-3105AEA 0 NOK
PROONIST-3105A
Beryllium standard solution
Standarder Kationstandarder Beryllium Standards
This reference standard solution is intended for use as a primary calibration standard for the quantitative determination of beryllium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of beryllium.
Sertifiseringer: Traceable to NIST and SRM numbers.