Chromium standard solution
Leverandør: LGC Standards PROMOCHEM
NIST-3112A
PROONIST-3112AEA 0 NOK
PROONIST-3112A
Chromium standard solution
Standarder Kationstandarder Chromium Standards
This reference standard solution is for use as a primary calibration standard for the quantitative determination of chromium.
One unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of chromium.
Sertifiseringer: Traceable to NIST and SRM numbers.