Indium standard solution
Leverandør: LGC Standards PROMOCHEM
NIST-3124A
PROONIST-3124AEA 0 NOK
PROONIST-3124A
Indium standard solution
Standarder Kationstandarder Indium Standards
This reference standard is intended for use as a primary calibration standard for the quantitative determination of indium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of indium.
Sertifiseringer: Traceable to NIST and SRM numbers.