Cyclosporin C ≥80%

Leverandør: LGC Standards PROMOCHEM
Danger

Synonymer: (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-33-((1R,2R,E)-1-Hydroxy-2-methylhex-4-en-1-yl)-30-((R)-1-hydroxyethyl)-6,9,18,24-tetraisobutyl-3,21-diisopropyl-1,4,7,10,12,15,19,25,28-nonamethyl-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecaone, Thr2-Cyclosporine

TRC-C988905-25
PROOTRC-C988905-25EA 0 NOK
PROOTRC-C988905-25
Cyclosporin C ≥80%
Cyclosporin C
Formel: C₆₂H₁₁₁N₁₁O₁₃
MW: 1218,6 g/mol
Smeltepunkt: 144,..1460 °C
Lagringstemperatur: Room Temperature
MDL nummer: MFCD06410945
CAS-nummer: 59787-61-0
New Product

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR