7-Epi Docetaxel

Leverandør: LGC Standards PROMOCHEM

TRCE585300-1MG
PROOTRCE585300-1MGEA 0 NOK
PROOTRCE585300-1MG
7-Epi Docetaxel
Standarder Organiske standarder
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
CAS-nr 153381-68-1
Formel C₄₃H₅₃NO₁₄
GHS-signal Danger
Fraktforhold RT
Lagringstemp −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR